Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mbd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010773.2 | NP_034903.2 | 4 - 5 | 1062 | 91 | |
GenBank mRNA | AF072243.1 | - | 4 - 5 | 1020 | 91 | |
AK147008.1 | - | 4 - 5 | 947 | 91 | ||
BC026698.1 | - | 4 - 5 | 1022 | 91 | ||
BC046607.1 | - | 4 - 5 | 1062 | 6399206 | 91 | |
BC057289.1 | - | 4 - 5 | 894 | 91 | ||
BC058864.1 | - | 4 - 5 | 882 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |