Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Matn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252563.1 | NP_001239492.1 | 4 - 5 | 876 | 73 | |
NM_013592.4 | NP_038620.2 | 4 - 5 | 1141 | 73 | ||
GenBank mRNA | AJ006140.1 | - | 4 - 5 | 860 | 73 | |
AK041073.1 | - | 4 - 5 | 849 | 73 | ||
AK160786.1 | - | 4 - 5 | 937 | 73 | ||
BC036558.1 | - | 4 - 5 | 876 | 1366191 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |