Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mbd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013594.2 | NP_038622.2 | 8 - 9 | 1041 | 72 | |
GenBank mRNA | AF072240.1 | - | 9 - 10 | 1020 | 72 | |
AK004624.2 | - | 8 - 9 | 1042 | 72 | ||
AK032535.1 | - | 5 - 6 | 539 | 72 | ||
AK156401.1 | - | 9 - 10 | 1032 | 72 | ||
AK166042.1 | - | 4 - 5 | 361 | 72 | ||
BC069837.1 | - | 8 - 9 | 1057 | 30435760 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |