Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Limd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013860.2 | NP_038888.2 | 1 - 2 | 1919 | 57 | |
GenBank mRNA | AJ132409.1 | - | 1 - 2 | 1908 | 57 | |
AK004806.1 | - | 1 - 2 | 1223 | 57 | ||
AK047198.1 | - | 1 - 2 | 1913 | 57 | ||
AK083970.1 | - | 1 - 2 | 1881 | 57 | ||
AK186032.1 | - | 1 - 2 | 100 | 57 | ||
BC056449.1 | - | 1 - 2 | 1794 | 5703026 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |