Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1200008822.mm.1 |
NM_021516.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008822.mm.1 |
NM_021516.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008822.mm.1 |
NM_022801.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008822.mm.1 |
NM_022801.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008822.mm.1 |
TC1200001241.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200008820.mm.1 |
NM_021516.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008820.mm.1 |
NM_021516.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008820.mm.1 |
NM_022801.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008820.mm.1 |
NM_022801.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008820.mm.1 |
TC1200001241.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200008819.mm.1 |
NM_021516.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008819.mm.1 |
NM_021516.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008819.mm.1 |
NM_022801.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008819.mm.1 |
NM_022801.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008819.mm.1 |
TC1200001241.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200008821.mm.1 |
NM_021516.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008821.mm.1 |
NM_021516.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008821.mm.1 |
NM_022801.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008821.mm.1 |
NM_022801.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008821.mm.1 |
TC1200001241.mm.1 |
MTA TC |