Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164885.1 | NP_001158357.1 | 4 - 5 | 827 | 86 | |
NM_022882.4 | NP_075020.2 | 4 - 5 | 703 | 86 | ||
GenBank mRNA | AF286723.1 | - | 4 - 5 | 703 | 86 | |
AK033662.1 | - | 4 - 5 | 740 | 86 | ||
AK086834.1 | - | 4 - 5 | 639 | 86 | ||
AK136472.1 | - | 1298 | 86 | |||
BC039698.1 | - | 4 - 5 | 816 | 5101211 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |