Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ggt7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144786.2 | NP_659035.2 | 11 - 12 | 1519 | 69 | |
GenBank mRNA | AK032051.1 | - | 11 - 12 | 1517 | 69 | |
AK131175.1 | - | 8 - 9 | 3542 | 69 | ||
BC005772.1 | - | 11 - 12 | 1500 | 3602314 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |