Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pycr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144795.3 | NP_659044.1 | 4 - 5 | 982 | 67 | |
GenBank mRNA | AK139185.1 | - | 4 - 5 | 983 | 67 | |
BC006727.1 | - | 5 - 6 | 627 | 3962093 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |