Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Utp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144826.3 | NP_659075.1 | 15 - 16 | 1459 | 80 | |
GenBank mRNA | AF334387.1 | - | 15 - 16 | 1786 | 80 | |
AK029151.1 | - | 15 - 16 | 1460 | 80 | ||
AK084976.1 | - | 15 - 16 | 1452 | 80 | ||
AK161178.1 | - | 4 - 5 | 653 | 80 | ||
AK164010.1 | - | 15 - 16 | 1465 | 80 | ||
AK169785.1 | - | 15 - 16 | 1466 | 80 | ||
BC018250.1 | - | 15 - 16 | 1423 | 4924739 | 80 | |
BC023920.1 | - | 15 - 16 | 1453 | 5323367 | 80 | |
BC024850.1 | - | 15 - 16 | 1454 | 5360921 | 80 | |
BC025074.1 | - | 3 - 4 | 336 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |