Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc17a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144836.2 | NP_659085.1 | 2 - 3 | 439 | 66 | |
GenBank mRNA | AK040717.1 | - | 2 - 3 | 440 | 66 | |
AK050184.1 | - | 2 - 3 | 422 | 66 | ||
AK050277.1 | - | 3 - 4 | 403 | 66 | ||
BC018306.1 | - | 1 - 2 | 77 | 4193755 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |