Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144847.1 | NP_659096.1 | 8 - 9 | 778 | 69 | |
GenBank mRNA | BC011468.1 | - | 8 - 9 | 711 | 4012880 | 69 |
BC012437.1 | - | 8 - 9 | 778 | 3710089 | 69 | |
BC025042.1 | - | 3 - 4 | 240 | 69 | ||
BC068117.1 | - | 13 - 14 | 1426 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |