Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smyd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144918.2 | NP_659167.2 | 9 - 10 | 913 | 70 | |
GenBank mRNA | AK158267.1 | - | 9 - 10 | 894 | 70 | |
AK162926.1 | - | 9 - 10 | 837 | 70 | ||
BC016525.1 | - | 9 - 10 | 900 | 3601356 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |