Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sez6l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252566.1 | NP_001239495.1 | 9 - 10 | 2077 | 69 | |
NM_001252567.1 | NP_001239496.1 | 9 - 10 | 1897 | 69 | ||
NM_144926.5 | NP_659175.1 | 9 - 10 | 2077 | 69 | ||
GenBank mRNA | AB206789.1 | - | 9 - 10 | 1678 | 69 | |
AK034088.1 | - | 9 - 10 | 2049 | 69 | ||
AK165738.1 | - | 9 - 10 | 2050 | 69 | ||
BC011475.1 | - | 9 - 10 | 1883 | 4191879 | 69 | |
BC079573.1 | - | 9 - 10 | 1466 | 5686330 | 69 | |
BC096615.1 | - | 9 - 10 | 1989 | 6467632 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |