Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2rx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145526.2 | NP_663501.2 | 2 - 3 | 793 | 107 | |
GenBank mRNA | AK140283.1 | - | 2 - 3 | 2061 | 107 | |
AK141847.1 | - | 2 - 3 | 458 | 107 | ||
BC023089.1 | - | 2 - 3 | 446 | 3984739 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |