Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc26a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145947.2 | NP_666059.2 | 7 - 8 | 1011 | 78 | |
GenBank mRNA | AF345194.1 | - | 6 - 7 | 913 | 78 | |
AK054327.1 | - | 7 - 8 | 1012 | 78 | ||
BC026928.1 | - | 7 - 8 | 990 | 4020760 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |