Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc45a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177628.2 | NP_001171099.1 | 3 - 4 | 1323 | 68 | |
NM_145977.2 | NP_666089.1 | 3 - 4 | 1404 | 68 | ||
GenBank mRNA | AK035428.1 | - | 3 - 4 | 1273 | 68 | |
AK135655.1 | - | 3 - 4 | 595 | 68 | ||
BC031381.1 | - | 3 - 4 | 1404 | 5050610 | 68 | |
BC034084.1 | - | 1 - 2 | 653 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |