Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdlim2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253736.1 | NP_001240665.1 | 4 - 5 | 454 | 86 | |
NM_145978.2 | NP_666090.1 | 4 - 5 | 381 | 86 | ||
GenBank mRNA | AK137991.1 | - | 3 - 4 | 437 | 86 | |
BC024556.1 | - | 4 - 5 | 381 | 4990983 | 86 | |
DV646822.1 | - | 4 - 5 | 354 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |