Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdm5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145997.2 | NP_666109.2 | 19 - 20 | 3413 | 98 | |
GenBank mRNA | AK134200.1 | - | 1 - 2 | 231 | 98 | |
AK135085.1 | - | 4 - 5 | 741 | 98 | ||
AK144877.1 | - | 19 - 20 | 3384 | 98 | ||
AK159079.1 | - | 18 - 19 | 3275 | 98 | ||
AK166055.1 | - | 19 - 20 | 3394 | 98 | ||
BC010717.1 | - | 18 - 19 | 2964 | 98 | ||
BC011506.3 | - | 19 - 20 | 3347 | 98 | ||
BC022581.1 | - | 4 - 5 | 688 | 98 | ||
BC080691.1 | - | 19 - 20 | 3099 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |