Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168679.1 | NP_001162150.1 | 2 - 3 | 235 | 84 | |
NM_001168680.1 | NP_001162151.1 | 2 - 3 | 271 | 84 | ||
NM_146010.2 | NP_666122.1 | 1 - 2 | 377 | 84 | ||
GenBank mRNA | AK077301.1 | - | 2 - 3 | 284 | 84 | |
BC025461.1 | - | 2 - 3 | 243 | 5344986 | 84 | |
BC027777.1 | - | 2 - 3 | 246 | 5351814 | 84 | |
BC029068.1 | - | 2 - 3 | 267 | 4947664 | 84 | |
BC029808.1 | - | 1 - 2 | 376 | 4947699 | 84 | |
BC034198.1 | - | 1 - 2 | 150 | 4986549 | 84 | |
DV060937.1 | - | 1 - 2 | 210 | 84 | ||
DV065030.1 | - | 2 - 3 | 412 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |