Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabrp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146017.3 | NP_666129.1 | 2 - 3 | 253 | 69 | |
GenBank mRNA | AK028946.1 | - | 2 - 3 | 253 | 69 | |
AK160577.1 | - | 2 - 3 | 257 | 69 | ||
BC023693.1 | - | 2 - 3 | 240 | 5344479 | 69 | |
BC025550.1 | - | 2 - 3 | 198 | 5320737 | 69 | |
BC027245.1 | - | 2 - 3 | 216 | 3602400 | 69 | |
BC031196.1 | - | 2 - 3 | 227 | 5059564 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Mammalian Gene Collection Mus musculus Probe spans exons |