Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Golph3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177669.1 | NP_001171140.1 | 2 - 3 | 336 | 59 | |
NM_001177670.1 | NP_001171141.1 | 3 - 4 | 525 | 59 | ||
NM_146133.3 | NP_666245.2 | 3 - 4 | 525 | 59 | ||
GenBank mRNA | AK160454.1 | - | 2 - 3 | 297 | 59 | |
AK167675.1 | - | 3 - 4 | 475 | 59 | ||
BC027194.1 | - | 3 - 4 | 435 | 4216692 | 59 | |
BC047147.1 | - | 3 - 4 | 500 | 6465983 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |