Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146150.2 | NP_666262.2 | 6 - 7 | 1099 | 100 | |
GenBank mRNA | AK031548.1 | - | 6 - 7 | 1099 | 100 | |
AK040377.1 | - | 5 - 6 | 1330 | 100 | ||
AK166470.1 | - | 6 - 7 | 1229 | 100 | ||
BC023786.1 | - | 5 - 6 | 477 | 100 | ||
BC026832.1 | - | 6 - 7 | 1149 | 4486176 | 100 | |
BC036128.1 | - | 6 - 7 | 1217 | 4971791 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Mus musculus Probe spans exons |