Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ipo13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146152.3 | NP_666264.1 | 4 - 5 | 1738 | 63 | |
GenBank mRNA | AK052257.1 | - | 4 - 5 | 1739 | 63 | |
AK149978.1 | - | 4 - 5 | 1721 | 63 | ||
BC031823.1 | - | 4 - 5 | 1137 | 4188768 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |