Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mybpc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146189.3 | NP_666301.2 | 15 - 16 | 1674 | 84 | |
GenBank mRNA | AK029282.1 | - | 15 - 16 | 1660 | 84 | |
AK132563.1 | - | 15 - 16 | 1664 | 84 | ||
BC029762.1 | - | 15 - 16 | 1644 | 3983351 | 84 | |
BC039184.1 | - | 15 - 16 | 1639 | 3980667 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |