Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Picalm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252520.1 | NP_001239449.1 | 8 - 9 | 1154 | 126 | |
NM_001252521.1 | NP_001239450.1 | 8 - 9 | 1154 | 126 | ||
NM_001252522.1 | NP_001239451.1 | 8 - 9 | 1154 | 126 | ||
NM_001252523.1 | NP_001239452.1 | 8 - 9 | 1154 | 126 | ||
NM_001252524.1 | NP_001239453.1 | 8 - 9 | 1154 | 126 | ||
NM_146194.4 | NP_666306.2 | 8 - 9 | 1154 | 126 | ||
GenBank mRNA | AK085472.2 | - | 5 - 6 | 388 | 126 | |
AK144960.1 | - | 8 - 9 | 893 | 127 | ||
AK162360.1 | - | 8 - 9 | 847 | 126 | ||
AK165066.1 | - | 3 - 4 | 265 | 126 | ||
AK170373.1 | - | 8 - 9 | 1174 | 126 | ||
AK220291.1 | - | 8 - 9 | 870 | 126 | ||
AY206701.1 | - | 8 - 9 | 838 | 126 | ||
BC011470.1 | - | 8 - 9 | 864 | 2651109 | 126 | |
BC021491.1 | - | 8 - 9 | 1103 | 5345558 | 126 | |
BC023843.1 | - | 8 - 9 | 902 | 5346041 | 126 | |
BC025566.1 | - | 8 - 9 | 1177 | 5322606 | 126 | |
BC027116.1 | - | 5 - 6 | 465 | 126 | ||
BC057683.1 | - | 8 - 9 | 1105 | 3671962 | 126 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |