Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc29a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146257.2 | NP_666369.1 | 5 - 6 | 748 | 57 | |
GenBank mRNA | BC025599.1 | - | 5 - 6 | 692 | 5250967 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |