Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500033747.mm.1 |
TC0500003694.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500033747.mm.1 |
NM_001163493.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500033747.mm.1 |
NM_001163493.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500033747.mm.1 |
NM_146258.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500033747.mm.1 |
NM_146258.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500033746.mm.1 |
TC0500003694.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500033746.mm.1 |
NM_001163493.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500033746.mm.1 |
NM_001163493.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500033746.mm.1 |
NM_146258.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500033746.mm.1 |
NM_146258.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500033745.mm.1 |
TC0500003694.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500033745.mm.1 |
NM_001163493.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500033745.mm.1 |
NM_001163493.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500033745.mm.1 |
NM_146258.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500033745.mm.1 |
NM_146258.2 |
Refseq |