Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fyco1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110253.2 | NP_001103723.2 | 7 - 8 | 850 | 111 | |
NM_148925.3 | NP_683727.3 | 8 - 9 | 1097 | 111 | ||
GenBank mRNA | AJ428065.1 | - | 8 - 9 | 1002 | 111 | |
AK081259.1 | - | 7 - 8 | 833 | 111 | ||
BC053712.1 | - | 6 - 7 | 758 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |