Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap1b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243043.1 | NP_001229972.1 | 18 - 19 | 2900 | 75 | |
NM_001243044.1 | NP_001229973.1 | 17 - 18 | 2682 | 75 | ||
NM_007454.3 | NP_031480.2 | 17 - 18 | 2682 | 75 | ||
GenBank mRNA | AA003413.1 | - | 3 - 4 | 318 | 75 | |
AK034140.1 | - | 18 - 19 | 2903 | 75 | ||
AK155892.1 | - | 17 - 18 | 2602 | 75 | ||
AK159273.1 | - | 17 - 18 | 2573 | 75 | ||
AK160042.1 | - | 17 - 18 | 2573 | 75 | ||
AK165747.1 | - | 17 - 18 | 2601 | 75 | ||
AK170376.1 | - | 17 - 18 | 2593 | 75 | ||
BC008513.1 | - | 17 - 18 | 2495 | 3601435 | 75 | |
BC066827.1 | - | 8 - 9 | 1226 | 75 | ||
Y07919.1 | - | 17 - 18 | 2537 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |