Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007462.3 | NP_031488.2 | 12 - 13 | 1852 | 79 | |
GenBank mRNA | AK046280.1 | - | 12 - 13 | 1842 | 79 | |
AK080907.1 | - | 12 - 13 | 1852 | 79 | ||
BC141141.1 | - | 11 - 12 | 1565 | 9055753 | 79 | |
M88127.1 | - | 11 - 12 | 1541 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |