Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gfap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010277.3 | NP_034407.2 | 7 - 8 | 1240 | 70 | |
GenBank mRNA | AF332061.1 | - | 7 - 8 | 1067 | 70 | |
AF332062.1 | - | 7 - 8 | 1067 | 70 | ||
AK079577.1 | - | 7 - 8 | 1175 | 70 | ||
BC100728.1 | - | 7 - 8 | 1119 | 70 | ||
BC100737.1 | - | 7 - 8 | 1112 | 70 | ||
BC101968.1 | - | 7 - 8 | 1119 | 70 | ||
BC103571.1 | - | 7 - 8 | 1113 | 70 | ||
BC139357.1 | - | 7 - 8 | 1240 | 8862379 | 70 | |
BC139358.1 | - | 7 - 8 | 1240 | 8862380 | 70 | |
K01347.1 | - | 7 - 8 | 1076 | 70 | ||
M25937.1 | - | 7 - 8 | 1076 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Mouse Stem Cell Pluripotency Inventoried Mouse_Stem_Cell Probe spans exons |