Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011763.2 | NP_035893.1 | 3 - 4 | 311 | 96 | |
GenBank mRNA | AK007823.1 | - | 3 - 4 | 290 | 96 | |
AK029052.1 | - | 3 - 4 | 297 | 96 | ||
BY132524.1 | - | 3 - 4 | 296 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons |