Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stk11ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027886.3 | NP_082162.3 | 17 - 18 | 2212 | 75 | |
GenBank mRNA | AK004757.2 | - | 17 - 18 | 2165 | 75 | |
AK156122.1 | - | 16 - 17 | 2300 | 75 | ||
AK165911.1 | - | 17 - 18 | 2204 | 75 | ||
AK171989.1 | - | 16 - 17 | 2074 | 75 | ||
AK195907.1 | - | 2 - 3 | 200 | 75 | ||
AK213443.1 | - | 1 - 2 | 118 | 75 | ||
BC035530.1 | - | 3 - 4 | 1252 | 75 | ||
BC042403.1 | - | 15 - 16 | 2249 | 75 | ||
BC061635.1 | - | 17 - 18 | 2195 | 6530178 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |