Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acsbg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053178.2 | NP_444408.1 | 6 - 7 | 947 | 65 | |
GenBank mRNA | AB050554.1 | - | 6 - 7 | 947 | 65 | |
AK054103.1 | - | 6 - 7 | 760 | 65 | ||
AK129179.1 | - | 6 - 7 | 743 | 65 | ||
BC057322.1 | - | 6 - 7 | 743 | 6832261 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |