Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdm4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172132.2 | NP_742144.1 | 6 - 7 | 880 | 120 | |
GenBank mRNA | AK129233.1 | - | 6 - 7 | 839 | 120 | |
AK141879.1 | - | 6 - 7 | 857 | 120 | ||
AK145151.1 | - | 6 - 7 | 832 | 120 | ||
BC007145.1 | - | 6 - 7 | 850 | 3490671 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |