Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mark4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172279.1 | NP_758483.1 | 2 - 3 | 254 | 68 | |
GenBank mRNA | AK146784.1 | - | 3 - 4 | 589 | 68 | |
AY151083.1 | - | 2 - 3 | 254 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mus musculus Probe spans exons |