Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172281.2 | NP_758485.2 | 5 - 6 | 834 | 103 | |
GenBank mRNA | AK089805.1 | - | 5 - 6 | 766 | 103 | |
AK129184.1 | - | 5 - 6 | 752 | 103 | ||
AK154982.1 | - | 5 - 6 | 765 | 103 | ||
AK171462.1 | - | 5 - 6 | 763 | 103 | ||
BC038348.1 | - | 5 - 6 | 748 | 4187484 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |