Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mab21l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172295.4 | NP_758499.3 | 5 - 6 | 1186 | 62 | |
GenBank mRNA | AK029040.1 | - | 5 - 6 | 925 | 62 | |
AK161057.1 | - | 5 - 6 | 1103 | 62 | ||
AK166928.1 | - | 936 | 62 | |||
AK172389.1 | - | 5 - 6 | 1135 | 62 | ||
BC037703.1 | - | 5 - 6 | 1123 | 4166596 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |