Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tex10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172304.3 | NP_758508.2 | 7 - 8 | 1883 | 97 | |
GenBank mRNA | AK132870.1 | - | 7 - 8 | 1884 | 97 | |
AK141288.1 | - | 6 - 7 | 1662 | 97 | ||
BC006867.1 | - | 4 - 5 | 1130 | 97 | ||
BC151176.1 | - | 6 - 7 | 1634 | 9088065 | 97 | |
BC151177.1 | - | 6 - 7 | 1634 | 9088066 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |