Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trak2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172406.3 | NP_765994.2 | 5 - 6 | 926 | 70 | |
GenBank mRNA | AK014647.1 | - | 5 - 6 | 875 | 70 | |
AK036650.1 | - | 5 - 6 | 875 | 70 | ||
BC060681.1 | - | 4 - 5 | 643 | 6410345 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |