Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trak2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172406.3 | NP_765994.2 | 6 - 7 | 1127 | 90 | |
GenBank mRNA | AK014647.1 | - | 6 - 7 | 1076 | 90 | |
AK036650.1 | - | 6 - 7 | 1076 | 90 | ||
BC060681.1 | - | 5 - 6 | 844 | 6410345 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |