Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc38a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172479.3 | NP_766067.2 | 6 - 7 | 543 | 98 | |
GenBank mRNA | AK054485.1 | - | 6 - 7 | 530 | 98 | |
AK077667.1 | - | 6 - 7 | 512 | 98 | ||
AK155928.1 | - | 6 - 7 | 496 | 98 | ||
AK158125.1 | - | 5 - 6 | 664 | 98 | ||
BC152401.1 | - | 6 - 7 | 512 | 40057282 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |