Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007713.4 | NP_031739.3 | 9 - 10 | 1831 | 82 | |
GenBank mRNA | AF033565.1 | - | 9 - 10 | 1107 | 82 | |
AK009908.1 | - | 5 - 6 | 550 | 82 | ||
AK090215.1 | - | 9 - 10 | 1072 | 82 | ||
AK133936.1 | - | 9 - 10 | 1077 | 82 | ||
AK136481.1 | - | 7 - 8 | 3470 | 82 | ||
AK140015.1 | - | 8 - 9 | 3174 | 82 | ||
AK145880.1 | - | 9 - 10 | 1086 | 82 | ||
AK146942.1 | - | 9 - 10 | 1834 | 82 | ||
AK154003.1 | - | 9 - 10 | 1080 | 82 | ||
AK167300.1 | - | 9 - 10 | 1833 | 82 | ||
BC085084.1 | - | 9 - 10 | 1074 | 6309020 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |