Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sin3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113248.2 | NP_001106719.1 | 3 - 4 | 400 | 68 | |
NM_009188.4 | NP_033214.2 | 3 - 4 | 400 | 68 | ||
GenBank mRNA | AF038848.1 | - | 3 - 4 | 380 | 68 | |
AK146847.1 | - | 3 - 4 | 383 | 68 | ||
AK159076.1 | - | 3 - 4 | 383 | 68 | ||
AK165592.1 | - | 3 - 4 | 385 | 68 | ||
AK169433.1 | - | 3 - 4 | 385 | 68 | ||
AK173005.1 | - | 3 - 4 | 369 | 68 | ||
BC020049.1 | - | 3 - 4 | 364 | 68 | ||
BC021160.1 | - | 3 - 4 | 383 | 68 | ||
BC051536.1 | - | 1 - 2 | 53 | 68 | ||
BC096367.1 | - | 3 - 4 | 368 | 68 | ||
L38622.2 | - | 3 - 4 | 372 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |