Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Araf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009703.2 | NP_033833.1 | 14 - 15 | 1683 | 56 | |
GenBank mRNA | AK034861.1 | - | 15 - 16 | 1790 | 56 | |
AK085823.1 | - | 14 - 15 | 1650 | 56 | ||
BC004757.1 | - | 14 - 15 | 1639 | 3154293 | 56 | |
D00024.1 | - | 9 - 10 | 1045 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |