Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srebf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011480.3 | NP_035610.1 | 14 - 15 | 2627 | 87 | |
GenBank mRNA | AF374266.1 | - | 13 - 14 | 2391 | 87 | |
AK052628.1 | - | 14 - 15 | 2628 | 87 | ||
AK150052.1 | - | 14 - 15 | 2616 | 87 | ||
AK154424.1 | - | 14 - 15 | 2595 | 87 | ||
AK158269.1 | - | 7 - 8 | 1176 | 87 | ||
AK169607.1 | - | 14 - 15 | 2628 | 87 | ||
BC006051.1 | - | 11 - 12 | 1763 | 87 | ||
BC056922.1 | - | 14 - 15 | 2609 | 6824948 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |