Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpt1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013495.2 | NP_038523.2 | 2 - 3 | 226 | 99 | |
GenBank mRNA | AF017175.1 | - | 114 | 99 | ||
AK050213.1 | - | 2 - 3 | 519 | 99 | ||
AK077454.1 | - | 2 - 3 | 180 | 99 | ||
AK128925.1 | - | 2 - 3 | 180 | 99 | ||
AK133472.1 | - | 2 - 3 | 181 | 99 | ||
AK136487.1 | - | 2 - 3 | 180 | 99 | ||
AK147770.1 | - | 2 - 3 | 181 | 99 | ||
AK149688.1 | - | 2 - 3 | 177 | 99 | ||
BC038395.1 | - | 1 - 2 | 161 | 2649352 | 99 | |
BC046383.1 | - | 1 - 2 | 161 | 4160676 | 99 | |
BC054791.1 | - | 2 - 3 | 207 | 6832731 | 99 | |
CJ081259.1 | - | 2 - 3 | 227 | 99 | ||
D49472.1 | - | 1 - 2 | 153 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |