Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpt1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013495.2 | NP_038523.2 | 6 - 7 | 779 | 93 | |
GenBank mRNA | AK050213.1 | - | 6 - 7 | 1072 | 93 | |
AK077454.1 | - | 6 - 7 | 733 | 93 | ||
AK128925.1 | - | 6 - 7 | 733 | 93 | ||
AK133472.1 | - | 6 - 7 | 734 | 93 | ||
AK136487.1 | - | 6 - 7 | 733 | 93 | ||
AK147770.1 | - | 6 - 7 | 734 | 93 | ||
AK149688.1 | - | 6 - 7 | 730 | 93 | ||
BC038395.1 | - | 5 - 6 | 714 | 2649352 | 93 | |
BC046383.1 | - | 5 - 6 | 714 | 4160676 | 93 | |
BC054791.1 | - | 6 - 7 | 760 | 6832731 | 93 | |
D49472.1 | - | 5 - 6 | 706 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |