Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpt1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013495.2 | NP_038523.2 | 9 - 10 | 1056 | 80 | |
GenBank mRNA | AF017175.1 | - | 944 | 80 | ||
AK050213.1 | - | 9 - 10 | 1349 | 80 | ||
AK077454.1 | - | 9 - 10 | 1010 | 80 | ||
AK128925.1 | - | 9 - 10 | 1010 | 80 | ||
AK133472.1 | - | 9 - 10 | 1011 | 80 | ||
AK136487.1 | - | 9 - 10 | 1010 | 80 | ||
AK147770.1 | - | 9 - 10 | 1011 | 80 | ||
AK149688.1 | - | 9 - 10 | 1007 | 80 | ||
BC038395.1 | - | 8 - 9 | 991 | 2649352 | 80 | |
BC046383.1 | - | 8 - 9 | 991 | 4160676 | 80 | |
BC054791.1 | - | 9 - 10 | 1037 | 6832731 | 80 | |
D49472.1 | - | 8 - 9 | 983 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |