Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exosc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027148.3 | NP_081424.3 | 5 - 6 | 580 | 69 | |
GenBank mRNA | AJ505005.1 | - | 5 - 6 | 240 | 69 | |
AK009584.1 | - | 5 - 6 | 252 | 69 | ||
AK169719.1 | - | 3 - 4 | 183 | 69 | ||
AK220227.1 | - | 5 - 6 | 580 | 69 | ||
BC059089.1 | - | 5 - 6 | 609 | 6509820 | 69 | |
BC067250.1 | - | 4 - 5 | 223 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |